N(α)-Fmoc-D-tryptophan
Supplier: Novabiochem (Part of Merck)
Certificates
About this item
Synonyms: Fmoc-D-Trp-OH
CAS Number:86123-11-7
:
Formula:C₂₆H₂₂N₂O₄
MDL Number:MFCD00062954
Molecular Weight:426.46 g/mol
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together