2-Methoxy-4-methyl-5-nitropyridine
Catalog # COBBOR-5581-5G
Supplier: COMBI-BLOCKS
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
- Environmentally Preferable:
Specifications
About this item
:
:168.15 g/mol
CAS Number:6635-90-1
Formula:C₇H₈N₂O₃
MDL Number:MFCD03095075