VWR® pHenomenal® MU 6100 L Multi-Parameter Meter (pH/Conductivity/DO), Bench
Supplier: VWR Collection
Certificates
About this item
User friendly instrument, with articulated electrode stand and IP 43-rated housing, provides high resolution and accuracy for precise multi-parameter measurements. This meter can either measure pH/mV and conductivity or pH/mV and oxygen simultaneously. Complete with USB output for connection to a computer. GLP compliant.
- Large graphic LCD with continuous LED backlight shows the measured parameter and the temperature value simultaneously
- Data logging function selectable between 1 min and 1 h; memory capacity of 5000 data sets
- Automatic power shut-off: Adjustable from 10 min to 24 h
- Automatic buffer recognition of DIN and NIST buffers (1,68 / 4,00 / 6,86 / 9,18 / 12,54)
- Three additional technical buffers at 25 °C (4,00 / 7,00 / 10,00) in memory
- Advanced self-test and diagnostics (electrode efficiency diagnostic, drift control); calibration symbol shows when recalibration is required
- Low energy consumption. Choice of mains supply or 4× AA batteries for over 1000 operating hours
Ordering information: Conductivity probes for low conductivities are available on request. Please contact your local Avantor sales office for further information.
Delivery: Ready to use MU 6100 L S1 kit (665-0309 ) includes bench multi-parameter meter, glass pH electrode pHenomenal 221 with built-in temperature sensor (1 m cable), conductivity probe CO 11 with built-in temperature sensor (1,5 m cable), articulated stand, 50 ml plastic beaker, 50 ml buffer 4, 50 ml buffer 7, plugin power supply, short instructions and instruction manual (22 languages) on CD ROM.
The MU 6100 L S2 kit (665-0310) includes meter, stand, 4 LR06 batteries, short instructions, CD ROM with manual, 50 ml plastic beaker, 50 ml buffer 4, 50 ml buffer 7, plugin power supply, pH electrode pHenomenal 221, 50 ml conductivity standard 0,01 mol/L KCl, conductivity cell CO 11, 30 ml cleaning solution, 50 ml electrolyte, 3 exchange caps, DO electrode OXY 11 (3 m cable).
The MU 6100 L S2 kit (665-0310) includes meter, stand, 4 LR06 batteries, short instructions, CD ROM with manual, 50 ml plastic beaker, 50 ml buffer 4, 50 ml buffer 7, plugin power supply, pH electrode pHenomenal 221, 50 ml conductivity standard 0,01 mol/L KCl, conductivity cell CO 11, 30 ml cleaning solution, 50 ml electrolyte, 3 exchange caps, DO electrode OXY 11 (3 m cable).
Specifications
- Catalog No:
- 665-0309
- 665-0310
- Model:
- MU 6100 L, S1 kit
- MU 6100 L, S2 kit
- pH range:
- −2,000...+19,999
- −2,000...+19,999
- pH resolution:
- 0,1/0,01/0,001 (selectable)
- 0,1/0,01/0,001 (selectable)
- pH accuracy:
- ±0,1/±0,01/±0,005
- ±0,1/±0,01/±0,005
- pH calibration:
- Up to 5 points
- Up to 5 points
- Automatic buffer recognition:
- VWR® Chemicals TITRINORM® buffers at 25 °C: 4,00/6,98/9,94
NIST/DIN 19266 (1,68/4,00/6,86/9,18/12,54)
Technical buffers at 25 °C: 4,00/7,00/10,00 - VWR® Chemicals TITRINORM® buffers at 25 °C: 4,00/6,98/9,94
NIST/DIN 19266 (1,68/4,00/6,86/9,18/12,54)
Technical buffers at 25 °C: 4,00/7,00/10,00
- mV range:
- ±1200,0/±2500
- ±1200,0/±2500
- mV resolution:
- 0,1/1
- 0,1/1
- mV accuracy:
- ±0,3/±1
- ±0,3/±1
- Conductivity range:
- 0,001 µS/cm - 1000 mS/cm
- 0,001 µS/cm - 1000 mS/cm
- Conductivity resolution:
- 0,1 µS/cm
- 0,1 µS/cm
- Conductivity accuracy:
- ±0,5% of measured value
- ±0,5% of measured value
- Conductivity cell constant:
- 0,001 - 25 cm⁻¹ (depending on probe)
- 0,001 - 25 cm⁻¹ (depending on probe)
- Conductivity calibration:
- 1 point
- 1 point
- Resistivity range:
- 1 Ω/cm - 999 MΩ/cm
- 1 Ω/cm - 999 MΩ/cm
- Resistivity accuracy:
- ±0,5% of measured value
- ±0,5% of measured value
- Salinity range:
- 0,00 - 70 ppt
- 0,00 - 70 ppt
- TDS range:
- 0 - 1999 mg/L / 2,00 - 19,99 g/L / 20,0 - 199,9 g/L
- 0 - 1999 mg/L / 2,00 - 19,99 g/L / 20,0 - 199,9 g/L
- TDS resolution:
- 1 mg/L / 0,01 g/L / 0,1 g/L
- 1 mg/L / 0,01 g/L / 0,1 g/L
- DO concentration:
- 0,00 - 20,00 / 0,0 - 90,0 mg/L
- 0,00 - 20,00 / 0,0 - 90,0 mg/L
- DO concentration resolution:
- 0,1%; 0,01 mg/L (depending on sensor)
- 0,1%; 0,01 mg/L (depending on sensor)
- DO saturation:
- 0,0 - 200,0; 0 - 600 % (depending on sensor)
- 0,0 - 200,0; 0 - 600 % (depending on sensor)
- DO accuracy:
- ±0,5% of value (between 5...30 °C)
- ±0,5% of value (between 5...30 °C)
- DO calibration:
- Saturated air
- Saturated air
- Temperature range:
- Auto: −5,0...+105,0 °C; manual: −25...+130 °C
- Auto: −5,0...+105,0 °C; manual: −25...+130 °C
- Temperature resolution:
- 0,1/1 °C
- 0,1/1 °C
- Temperature accuracy:
- ±0,2 °C
- ±0,2 °C
- Temperature compensation:
- Automatic/manual
- Automatic/manual
- Display:
- Graphic LCD with LED backlight
- Graphic LCD with LED backlight
- Power supply:
- Mains supply or 4× AA batteries
- Mains supply or 4× AA batteries
- Compliance:
- IP 43/GLP
- IP 43/GLP
- Data logging - log function:
- Selectable between 1 s and 1 h (selectable intervals: 1 s, 5 s, 10 s, 30 s, 1 min, 5 min, 10 min, 15 min, 30 min, 60 min)
- Selectable between 1 s and 1 h (selectable intervals: 1 s, 5 s, 10 s, 30 s, 1 min, 5 min, 10 min, 15 min, 30 min, 60 min)
- Data logging capacity:
- 5000 data sets
- 5000 data sets
- Connections:
- BNC for pH/banana for temperature/8-pin for conductivity and DO
- BNC for pH/banana for temperature/8-pin for conductivity and DO
- Outputs:
- Waterproof USB connection (slave, bidirectional)
- Waterproof USB connection (slave, bidirectional)
Specifications
pH electrode
Product Information
Frequently Bought Together