Reversed phase HPLC columns, ProSwift™ RP
Reversed phase columns provide the advantages of high resolution at high flow rates for fast protein separation and analysis.
- High resolution at high speed
- Operational flow rates available
- Optimal performance in a board range of applications
- pH range 1 to 4
- High loading capacity
Frequently Bought Together