Vacuum manifold and accessories
The multi-well plate vacuum manifold is an anodised aluminium manifold that has been designed and optimised for the vacuum filtration of AcroPrep™, AcroPrep Advance, and AcroWell™ multi-well filter plates.
- Supplied complete with the necessary O-ring and gasket; control block includes vacuum pressure gauge, vacuum metering valve, vacuum release valve, and ¼" hose barb for vacuum line attachment
- Vacuum manifold unit includes a Delrin® plastic spacer block designed to accommodate standard 350 μl receiver plates and optimised to reduce space between the receiver plate and the filter plate during vacuum filtration
- Optional spacer block available for use with receiver plates
- Adapter collar holds filter plates tightly to receiver plates for centrifugation
Frequently Bought Together