Order Entry
United Kingdom
ContactUsLinkComponent
Protein purification system, ÄKTA™ start
Protein purification system, ÄKTA™ start
Catalog # 29-0220-94
Supplier:  Cytiva
CAS Number:  
undefined
Protein purification system, ÄKTA™ start
Catalog # 29-0220-94
Supplier:  Cytiva
CAS Number:  

Specifications

  • Pk:
    1
  • Description:
    ÄKTA™ start, main instrument
  • Seq:
    02
  • Environmentally Preferable:
  • Model:
    ÄKTA™ start
  • Flow cell path length:
    2 mm optical path length, 2 µl cell volume (default)
  • Viscosity:
    0,6 - 5 cP
  • Operating pressure:
    0,5 Mpa max
  • UV wavelength:
    280 nm
  • Application:
    Entry level chromatography solution for the preparative preparation of proteins on laboratory scale
  • Flow rate:
    0,5 - 5 ml/min
  • Inlet/outlet connections:
    3 ports per valve (buffer valve and sample valve: 2 in/1 out; wash valve and outlet valve: 1 in/2 out)
  • Power consumption:
    95 VA
  • Protection class:
    IP 21
  • Pump:
    Peristaltic pump; single channel, 4 roller pump head with low pulsation
  • Voltage:
    220 - 240 VAC
  • Weight:
    9 kg
  • W×D×H:
    340×280×360 mm

Specifications

About this item

ÄKTA start is an easy to use chromatography system that automates manual purification procedures using, for example, HiTrap columns. It offers an entry level chromatography solution for the preparative purification of proteins at lab scale.

  • Intuitive touch screen display and real time monitoring
  • Compact solution for quick and reliable one-step protein purification
  • Quick start methods and templates for common purification techniques
  • Sample injection and fraction collection options

The system is easy to use and has an intuitive design, front-facing work area, visible flow path, and a user friendly interface. ÄKTA™ start makes it easy to purify proteins with pre-defined quick start and editable method templates, real time run control and visualisation, and single-click operations, together with a compact and portable size suitable for cold room operations. This instrument can be controlled either in stand-alone mode, using the instrument's touch screen display, or using UNICORN™ start control software on a computer.

ÄKTA™ start supports commonly used chromatography techniques like desalting/buffer exchange, affinity, gel filtration, and ion exchange. Using ÄKTA™ start you can perform typical applications such as purification of affinity-tagged proteins, antibodies, untagged or native proteins, and sample clean-up.

Frac30, a round fraction collector, is controlled via the ÄKTA™ start touch screen instrument display or using UNICORN™ start software on a computer. Frac30 allows you to collect up to 30 fractions and supports four tube sizes, ranging from 1,5 to 15 ml. Fractions can be collected in volumes ranging from 0,5 to 15 ml. The entire fraction collector rack can be transferred as required.

UNICORN™ system control software, especially designed for ÄKTA™ start, includes a process picture map for real time monitoring, an intuitive method editor, and evaluation modules.

Certifications: CE compliant.
Ordering information: Main instrument includes stand-alone system with accessory box and user documentation. Fraction collector (Frac30) and UNICORN™ start software are optional modules. Please contact your local Avantor sales office for further information.

Optional modules

Product Information