Tissue culture tubes, Leighton type
Supplier: BELLCO GLASS
Certificates
About this item
Glass, clear.
Leighton culture tubes are useful for the study of pathogenic effects on viruses on cells as well as sponge matrix culture and nutritional studies. Use cover slips for preparing explant tissue cultures with Leighton culture tubes.
Frequently Bought Together