Barium chloride 250 g/l in aqueous solution
Catalog # PRLS88384.360
Supplier: VWR Collection
CAS Number:
Specifications
- Pk:5 L
- Seq:5190
- Environmentally Preferable:
Specifications
About this item
:
:208.24 g/mol
EINECS:233-788-1
UN:3287
:GHS07
Storage Temperature:Ambient
CAS Number:10361-37-2
Formula:BaCl₂
MDL Number:MFCD00003445
Boiling Pt:∼100 °C (1013 hPa)
Melting Pt:963
ADR:6.1 III
Merck Index:12,00998