Citric acid, anhydrous ≥99.8%, powder, AnalaR NORMAPUR®
84841.290
:
- Pack type:Plastic bottle for solids
- Pk:1 kg
- Seq:4024
- Environmentally Preferable:
- Assay:Min. 99.8 %
- Ignition residue (SO4):Passes test USP
- Heavy metals (as Pb):Max. 1 ppm
- C2O4 (as C2H2O4):Max. 10 ppm
- Cl (Chloride):Max. 5 ppm
- SO4 (Sulphate):Max. 30 ppm
- Al (Aluminium):Max. 0.2 ppm
- As (Arsenic):Max. 1 ppm
- Ba (Barium):Max. 1 ppm
- Ca (Calcium):Max. 20 ppm
- Cu (Copper):Max. 1 ppm
- Fe (Iron):Max. 1 ppm
- Hg (Mercury):Max. 1 ppm
- Mg (Magnesium):Max. 1 ppm
- Pb (Lead):Max. 0.5 ppm
- Zn (Zinc):Max. 1 ppm
: 2-Hydroxypropane-1,2,3-tricarboxylic acid, Hydroxytricarballylic acid
:
:192.13 g/mol
:310
:201-069-1
:01-2119457026-42
:GHS07
:Ambient
:77-92-9
:HOC(COOH)(CH₂COOH)₂
:MFCD00011669
:149…151 °C
:1.665 g/cm³ (18 °C. OECD 109)
:13,02350