4-tert-Butylbenzamide ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:56108-12-4
:
Formula:C₁₁H₁₅NO
MDL Number:MFCD00017139
Molecular Weight:177.25 g/mol
Melting Pt:171…175 °C
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together