Potassium thiocyanate 8 mol/l in aqueous solution, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: Potassium rhodanide
CAS Number:333-20-0
:
Formula:KSCN
MDL Number:MFCD00011413
Boiling Pt:500
Molecular Weight:97.18 g/mol
Melting Pt:175
Merck Index:12,07866
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:206-370-1
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together