Diethylzinc 15% (w/w) 1 M in hexane, AcroSeal®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:557-20-0
:
Formula:C₄H₁₀Zn
MDL Number:MFCD00009021
Molecular Weight:123.51 g/mol
Melting Pt:39…28 °C
Flash Pt:–40 °C
Storage Temperature:Ambient
EINECS:209-161-3
UN:3394
ADR:4.2
:GHS07
:GHS08
:GHS05
:GHS09
:GHS02
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Clear to hazy colorless to yellow to grey-brown Solution
- Titration Complexometric:0.85 to 1.00 M
Specification Test Results
Related Information
Frequently Bought Together