Cyclopropylmagnesium bromide 0.5 M in MeTHF (2-methyltetrahydrofuran)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:23719-80-4
:
Formula:C₃H₅BrMg
MDL Number:MFCD00672094
Molecular Weight:145.28 g/mol
UN:3399
ADR:4.3 II
:GHS02
:GHS05
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together