Flame-retardant fleece jackets, Obaix 7759
: SIOEN
Jacket made of double-sided, flame-retardant polyester with anti-pilling treatment and stitched seams. Can be zipped into different jackets.
- Good cold protection, breathable
- Straight collar, zip closure with inside flap
- Two inset pockets with zip closure, one inside pocket
- Round inset sleeves
- Elastic drawstring in hem
Weight: 275 g/m²
: EN ISO 14116 (Index 1), EN 14058 (1XXXX)
- Catalog No:
- SIOE7759/B75/S
- SIOE7759/B75/M
- SIOE7759/B75/L
- SIOE7759/B75/3XL
- SIOE7759/B75/2XL
- SIOE7759/B75/1XL
- Model:
- Obaix 7759
- Obaix 7759
- Obaix 7759
- Obaix 7759
- Obaix 7759
- Obaix 7759
- Garment application:
- Fire protection
- Fire protection
- Fire protection
- Fire protection
- Fire protection
- Fire protection
- Risk area:
- Flame-retardant, Cold
- Flame-retardant, Cold
- Flame-retardant, Cold
- Flame-retardant, Cold
- Flame-retardant, Cold
- Flame-retardant, Cold
- Gender:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Closure type:
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Fabric:
- Polyester
- Polyester
- Polyester
- Polyester
- Polyester
- Polyester
- Weight:
- 275 g/m²
- 275 g/m²
- 275 g/m²
- 275 g/m²
- 275 g/m²
- 275 g/m²
Frequently Bought Together