Hydrogen peroxide OXYBIO L12
Catalog # VWRS5854.1006
Supplier: VWR Collection
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:1 L
- Seq:5037
- Environmentally Preferable:
Specifications
About this item
:
:34.01 g/mol
Boiling Pt:150.2
EINECS:231-765-0
UN:2984
:GHS05
Storage Temperature:Ambient
CAS Number:7722-84-1
Formula:H₂O₂
MDL Number:MFCD00011333
Melting Pt:–33 °C
Density:1.040 g/cm³ (20 °C)
ADR:5.1 III
Merck Index:14,04798