HPLC tubing, Tefzel® (ETFE)
Supplier: UPCHURCH SCIENTIFIC
Certificates
About this item
Tefzel® (ETFE), natural.
- Excellent solvent resistance
- More durable and less gas permeable than PTFE, FEP and PFA
- Operating temperatures up to 80 °C
ETFE tubing is chemically inert and suitable for higher pressure applications (when using aqueous mobile phase) than PTFE, FEP and PFA. Additionally, because Tefzel® is more rigid than PTFE, FEP and PFA, this tubing better resists inner diameter collapse.
¹/₁₆" ext. Ø tubing holds from 500 psi (34 bar) to 4000 psi (276 bar), depending upon the inner diameter; inner and outer diameter tolerances are ±0,001".
¹/₈" ext. Ø tubing holds from 500 psi (34 bar) to 1000 psi (69 bar), depending upon the inner diameter; inner and outer diameter tolerances are ±0,003".
Frequently Bought Together