Laboratory jacks, Swiss Boy
Supplier: GRAUER RUDOLF
Certificates
About this item
These laboratory jacks are made of anodised aluminium for corrosion resistance and resistance to many chemicals.
- Easy adjustment with resilience
- High stability and high load carrying capacity
The motorised models type 116 and 122 feature a variable adjustment of the lower and upper end positions. When the set position has been reached the lab jack stops automatically. These models are also suitable for repetitive operations.
Frequently Bought Together