Order Entry
United Kingdom
ContactUsLinkComponent
Chemical spill neutralisers, Ampho-Mag™
Chemical spill neutralisers, Ampho-Mag™
Catalog # NEWPPLPE218-IT
Supplier:  NEW PIG
Chemical spill neutralisers, Ampho-Mag™
Catalog # NEWPPLPE218-IT
Supplier:  NEW PIG

Specifications

  • Description:
    Neutralisers, Ampho-Mag™ (white) for acidic or caustic spills
  • Seq:
    02
  • Environmentally Preferable:
  • Packed:
    18,1 kg/container
  • Pk:
    18,1 kg

Specifications

About this item

Non toxic amphoteric neutralisers with colour pH indicator.

  • Colour pH indicator: red for acid/blue for alkali
  • Safely neutralised chemical spills turn to greenish yellow
  • Non abrasive composition will not harm floors or expensive laboratory countertops
Packaging: In an easy to use, light weight shaker bottle (1,4 kg).