β-D-Allose, Sigma-Aldrich®
Catalog # SIALA6390-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
- Environmentally Preferable:
Specifications
About this item
Synonyms: (2R,3R,4R,5S,6R)-6-(hydroxymethyl)oxane-2,3,4,5-tetrol, β-D-Allopyranose, (2R,3R,4R,5S,6R)-6-(Hydroxymethyl)tetrahydro-2H-pyran-2,3,4,5-tetraol
:
:180.16 g/mol
CAS Number:7283-09-2
Formula:C₆H₁₂O₆
MDL Number:MFCD00063268
:M-Clarity™ quality level = MQ200