D-(+)-Glucose-[¹³C₆] (99% ¹³C ), Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: (2R,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal-1,2,3,4,5,6-13C6, D-Glucose-13C6
CAS Number:110187-42-3
:
Formula:C₆H₁₂O₆
MDL Number:MFCD00037544
Molecular Weight:186.110 g/mol
:M-Clarity™ quality level = MQ200
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together