E.Z.N.A.® miRNA isolation kit
Supplier: OMEGA BIO-TEK
Certificates
About this item
The E.Z.N.A.® miRNA isolation kit uses a rapid procedure to isolate small RNAs, such as micro RNA (miRNA), small interfering RNA (siRNA), and small nuclear RNA (snRNA), from wide range of tissues and cells. The fast and efficient silica membrane based method isolates total RNA ranging in size from kilo based down to decamers. The kit also provides reagents and a procedure to enrich the population of RNAs that are 200 bases and smaller, which enhances the sensitivity of small RNA detection by solution hybridisation, Northern analysis, and other methods.
- Efficient isolation of small RNA containing total RNA
- Enrich for small RNA <200 nt to increase sensitivity in downstream analysis
- Simple 30 minutes procedure
- Compatible with virtually all cell and tissue type
Specifications
- Application:
- Method / Format:
- Refine target molecule:
- Sample type:
- Target molecule:
- Environmentally Preferable:
Specifications
Frequently Bought Together