Order Entry
United Kingdom
ContactUsLinkComponent
VWR® ESD Overalls, with Hood
undefined
VWR® ESD Overalls, with Hood
  • Catalog No:
  • 113-8308
  • 113-8309
  • 113-8310
  • 113-8305
  • 113-8306
  • 113-8307
  • Anti-static:
  • +
  • +
  • +
  • +
  • +
  • +
  • Cleanroom class:
  • 100/ISO 5
  • 100/ISO 5
  • 100/ISO 5
  • 100/ISO 5
  • 100/ISO 5
  • 100/ISO 5
  • Garment application:
  • General Purpose, Industrial
  • General Purpose, Industrial
  • General Purpose, Industrial
  • General Purpose, Industrial
  • General Purpose, Industrial
  • General Purpose, Industrial
  • Weight:
  • 50 g/m²
  • 50 g/m²
  • 50 g/m²
  • 50 g/m²
  • 50 g/m²
  • 50 g/m²
Frequently Bought Together