N-Phenylglycine 93%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 2-Anilinoacetic acid, 2-(Phenylamino)acetic acid
CAS Number:103-01-5
:
Formula:C₈H₉NO₂
MDL Number:MFCD00014009
Molecular Weight:151.17 g/mol
Melting Pt:121…127 °C
Storage Temperature:Ambient
EINECS:203-070-2
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Ochre to yellow-brown Powder
- Infrared spectrum:Conforms
- Melting point:121 to 127 °C
- HPLC:≥91 %
- Titration with TBAH:≥91 %
Specification Test Results
Frequently Bought Together