Order Entry
United Kingdom
ContactUsLinkComponent
E.Z.N.A.® and E-Z 96® Cycle-Pure Kits
E.Z.N.A.® and E-Z 96® Cycle-Pure Kits
Supplier:  OMEGA BIO-TEK
Certificates
undefined
E.Z.N.A.® and E-Z 96® Cycle-Pure Kits
Supplier:  OMEGA BIO-TEK
Certificates

Specifications

  • Application:
  • Method / Format:
  • Refine target molecule:
  • Sample type:
  • Target molecule:
  • Environmentally Preferable:

Specifications

Frequently Bought Together