Order Entry
United Kingdom
ContactUsLinkComponent
Dirt and stain remover, Miraclean
Dirt and stain remover, Miraclean
Catalog # 129-0991
Supplier:  FHP VILEDA SCS
undefined
Dirt and stain remover, Miraclean
Catalog # 129-0991
Supplier:  FHP VILEDA SCS
Specifications
  • Colour:
    White
  • Description:
    Miraclean dirt remover
  • L×W:
    120×75 mm
  • Environmentally Preferable:
  • Pk:
    8
Specifications
About this item
This dirt and stain remover made from 100% melamine resin can be used for removing tough marks, stains and ground-in dirt.
  • More efficient than any microfibre cleaning cloth
  • No chemicals needed, works with water only