Chemiluminescent detection reagents, Immobilon® Western HRP substrates
: Merck
Immobilon® Western HRP substrates are compatible with both PVDF and nitrocellulose membranes, as well as all commonly used buffers and blocking reagents.
- Environmentally Preferable:
Frequently Bought Together