Order Entry
United Kingdom
ContactUsLinkComponent
Affinity chromatography columns, HiTrap™ IMAC HP
Affinity chromatography columns, HiTrap™ IMAC HP
Supplier:  Cytiva
Certificates
undefined
Affinity chromatography columns, HiTrap™ IMAC HP
Supplier:  Cytiva

Specifications

  • Catalog No:
  • 17-0920-03
  • 17-0920-05
  • Max. pressure:
  • 5 bar
  • 5 bar
  • pH range:
  • 3 - 12
  • 3 - 12
  • Storage:
  • 4 to 30 °C, 20% Ethanol
  • 4 to 30 °C, 20% Ethanol

Specifications

Frequently Bought Together