Order Entry
United Kingdom
ContactUsLinkComponent
Affinity chromatography columns, HiTrap™ Protein A HP
Affinity chromatography columns, HiTrap™ Protein A HP
Supplier:  Cytiva
Certificates
undefined
Affinity chromatography columns, HiTrap™ Protein A HP
Supplier:  Cytiva
Certificates

Specifications

  • Catalog No:
  • 29-0485-76
  • 17-0403-03
  • 17-0402-01
  • 17-0403-01
  • 17-0402-03
  • Max. pressure:
  • 5 bar
  • 5 bar
  • 5 bar
  • 5 bar
  • 5 bar
  • pH range:
  • 3 - 9
  • 3 - 9
  • 3 - 9
  • 3 - 9
  • 3 - 9
  • Storage:
  • 4 to 8 °C, 20% Ethanol
  • 4 to 8 °C, 20% Ethanol
  • 4 to 8 °C, 20% Ethanol
  • 4 to 8 °C, 20% Ethanol
  • 4 to 8 °C, 20% Ethanol

Specifications

Frequently Bought Together