1-Boc-4-nitroindole
Supplier: ACTIVATE SCIENTIFIC
Certificates
About this item
Synonyms: 4-Nitroindole, N-BOC protected, tert-Butyl 4-nitro-1H-indole-1-carboxylate
CAS Number:913836-24-5
:
Formula:C₁₃H₁₄N₂O₄
MDL Number:MFCD08689543
Molecular Weight:262.27 g/mol
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together