4-(Trifluoromethoxy)phenylacetonitrile
Supplier: ACTIVATE SCIENTIFIC
Certificates
About this item
Synonyms: 1-(Cyanmethyl)-4-(trifluormethoxy)benzol, 4-(TrifluoroMethoxy)phenylacetonitrile, 4-(Trifluoromethoxy)benzyl cyanide
CAS Number:49561-96-8
:
Formula:C₉H₆F₃NO
MDL Number:MFCD00061240
Boiling Pt:225
Melting Pt:28…30 °C
Density:1.28 g/cm³ (20 °C)
Molecular Weight:201.15
UN:3439
ADR:6.1 III
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together