5-Bromo-2-iodopyrimidine
Supplier: ACTIVATE SCIENTIFIC
Certificates
About this item
CAS Number:183438-24-6
:
Formula:C₄H₂BrIN₂
MDL Number:MFCD01318111
Molecular Weight:284.88 g/mol
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together
2-Propanol ≥99.7%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Methanol ≥99.8%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Ethanol ≥96% (v/v), TechniSolv®, pure
Supplier: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Toluene ≥98%, TECHNICAL
Supplier: VWR Chemicals
Water Ph. Eur., USP, JP, purified
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
5-Bromo-2-iodopyrimidine
Supplier: Apollo Scientific
5-Hexyn-1-ol, Sigma-Aldrich®
Supplier: Merck