1-Bromo-4-hexylbenzene
Supplier: ACTIVATE SCIENTIFIC
Certificates
About this item
Synonyms: (4-Bromophenyl)hexane
CAS Number:23703-22-2
:
Formula:C₁₂H₁₇Br
MDL Number:MFCD00061114
Molecular Weight:241.17 g/mol
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together