3-(3,4-Methylenedioxyphenyl)propionic acid
Catalog # FLUO018840-5G
Supplier: FLUOROCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
- Environmentally Preferable:
Specifications
About this item
Synonyms: 3,4-(Methylenedioxy)hydrocinnamic acid, 3-(1,3-Benzodioxyl-5-yl)propionic acid
:
:194.19 g/mol
EINECS:220-565-9
CAS Number:2815-95-4
Formula:C₁₀H₁₀O₄
MDL Number:MFCD00016909