Order Entry
United Kingdom
ContactUsLinkComponent
 
2,2-Dimethoxy-2-phenylacetophenone 99%
 
undefined
2,2-Dimethoxy-2-phenylacetophenone 99%

 

  • Environmentally Preferable:

 

 

  • Appearance:
    White to light yellow Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    64 to 68 °C
  • Loss on drying:
    ≤0.2 % (75 °C, 2 hrs)
  • Heavy metals:
    ≤50 ppm
  • GC:
    ≥98.5 %

 

 

Frequently Bought Together