Hydrogen chloride 1 M in ethyl acetate
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7647-01-0
:
Formula:HCl
MDL Number:MFCD00011324
Density:0.900
Molecular Weight:36.46 g/mol
Storage Temperature:Refrigerator
EINECS:231-595-7
UN:2924
ADR:3 II
:GHS02
:GHS05
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together