3-Biphenylboronic acid
Supplier: FLUOROCHEM
Certificates
About this item
Synonyms: [1,1'-Biphenyl]-3-ylboronic acid
CAS Number:5122-95-2
:
Formula:C₁₂H₁₁BO₂
MDL Number:MFCD01318102
Molecular Weight:198.03 g/mol
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together