Order Entry
United Kingdom
ContactUsLinkComponent
Handheld airborne particle counters, MET One HHPC
Handheld airborne particle counters, MET One HHPC
Supplier:  Beckman Coulter
Certificates
undefined
Handheld airborne particle counters, MET One HHPC
Supplier:  Beckman Coulter
Specifications
  • Catalog No:
  • 710-0979
  • 710-0982
  • 710-0980
  • Model:
  • MET One HHPC 3+, 3 channels (0,3 - 10 µm)
  • MET One HHPC 6+, 6 channels (0,3 - 10 µm)
  • MET One HHPC 2+, 2 channels (0,5 - 5 µm)
  • Flow rate:
  • 2,83 L/min, 0,3 µm/min
  • 2,83 L/min, 0,3 µm/min
  • 2,83 L/min, 0,5 µm/min
  • Material:
  • Polycarbonate
  • Polycarbonate
  • Polycarbonate
  • Power:
  • 110/240 V
  • 110/240 V
  • 110/240 V