Order Entry
United Kingdom
ContactUsLinkComponent
PP1 solution (Acetonitrile: Formic Acid 96% (100:5 v/v))
PP1 solution (Acetonitrile: Formic Acid 96% (100:5 v/v))
Catalog # 799562E
Supplier:  VWR Collection
CAS Number:  
PP1 solution (Acetonitrile: Formic Acid 96% (100:5 v/v))
Catalog # 799562E
Supplier:  VWR Collection
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440
  • Environmentally Preferable:

Specifications

About this item

 :
UN:2924
 :GHS02
 :GHS07
Storage Temperature:Ambient
ADR:3 II