Ethyl-2-oxocyclohexanecarboxylate 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Ethyl 2-oxocyclohexanecarboxylate
CAS Number:1655-07-8
:
Formula:C₉H₁₄O₃
MDL Number:MFCD00001631
Boiling Pt:106 °C (11 mmHg)
Density:1.064
Molecular Weight:170.21 g/mol
Flash Pt:96 °C (205 °F)
Storage Temperature:Ambient
EINECS:216-731-5
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together