Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United Kingdom
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-(Methylthio)benzothiazole
2-(Methylthio)benzothiazole
2-(Methylthio)benzothiazole
EHERC15144200
:
EHRENSTORFER
:
2-(Methylthio)benzothiazole
EHERC15144200
:
EHRENSTORFER
:
Pack type:
Glass bottle for solids
Pk:
0,1 g
Seq:
3020
Environmentally Preferable:
:
2-(Methylthio)benzo[d]thiazole
:
:
181.2779 g/mol
:
210-417-1
:
615-22-5
:
C₈H₇NS₂
:
MFCD00005784