Order Entry
United Kingdom
ContactUsLinkComponent
Dimethylethoxysilane 94%
Dimethylethoxysilane 94%
Certificates
undefined
Dimethylethoxysilane 94%

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.3670 to 1.3700 (20°C, 589 nm)
  • GC:
    ≥92 %

Specification Test Results

Related Information

Frequently Bought Together