Millicell® Cell Culture Inserts, PC Membrane (Isopore™)
: Merck
With Millicell inserts, attachment or suspension cells can access media from both their apical and basolateral sides. Cell growth, structure, and function more closely mimic what occurs in vivo. In addition, Millicell inserts make it possible to study both sides of the cell monolayer.
The inserts are easily prepared for SEM and TEM visualising techniques, and they are compatible with cellular and/or fluorescent stains.
- Catalog No:
- MILFPIXP01250
- MILFPIHP01250
- MILFPITP01250
- MILFPIHP03050
- MILFPI8P01250
- Effective diameter of membrane:
- 12 mm
- 12 mm
- 12 mm
- 30 mm
- 12 mm
- Overall height:
- 10,5 mm
- 10,5 mm
- 10,5 mm
- 13 mm
- 10,5 mm
Frequently Bought Together