Order Entry
United Kingdom
ContactUsLinkComponent
Measuring beakers with handle
Measuring beakers with handle
Catalog # VITL44091
Supplier:  VITLAB
Supplier #: 44091
undefined
Measuring beakers with handle
Catalog # VITL44091
Supplier:  VITLAB
Supplier #: 44091
Log in for availability
Select an Option
70 mm
Select an Option
5 ml
Select an Option
250 ml
Select an Option
120 mm
Select an Option
01
Select an Option
1

About this item

SAN, crystal clear.

  • Temperature resistance up to 70 °C
  • With a raised scale
  • Stable, easy-grip handle

Ideal for general laboratory use and a wide variety of other applications. With embossed scale.

Frequently Bought Together