Cuvettes for spectrophotometers
Extensive range of cuvettes for spectrophotometry for measuring liquids.
Glass cuvettes are designed for the visible region whereas quartz cuvettes may be used in both the UV and visible regions.
Frequently Bought Together