Order Entry
Slovakia
ContactUsLinkComponent
1-Undecene
1-Undecene
Catalog # EHERC17896700
Supplier:  EHRENSTORFER
CAS Number:  
1-Undecene
Catalog # EHERC17896700
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,1 g
  • Seq:
    3020
  • Environmentally Preferable:

Specifications

About this item

 :
 :154.3 g/mol
Boiling Pt:193
Melting Pt:–48.9 °C
Density:0.7552 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:212-483-7
CAS Number:821-95-4
Formula:C₁₁H₂₂
MDL Number:MFCD00008956