2-Chloro-2-methylpropane ≥98.0% (by GC)
TCIAC0208-500ML
: TCI
:
- Pack type:Glass bottle
- Pk:500 mL
- Seq:2725
- Environmentally Preferable:
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
: tert-Butyl chloride
:
:92.57 g/mol
:208-066-4
:1127
:GHS02
:Refrigerator
:507-20-0
:C₄H₉Cl
:MFCD00000816
:52°C
:-28
:–18°C
:3 II
:14,01562