Order Entry
Slovakia
ContactUsLinkComponent
Pipette controller, Midi Plus™
Pipette controller, Midi Plus™
Catalog # 612-2715
Supplier:  SARTORIUS BIOHIT
undefined
Pipette controller, Midi Plus™
Catalog # 612-2715
Supplier:  SARTORIUS BIOHIT

Specifications

  • Description:
    Pipette controller Midi Plus, EU plug
  • Environmentally Preferable:
  • Pk:
    1

Specifications

About this item

Rechargeable, cordless and lightweight, the Midi Plus™ pipette controller can be used with all plastic, glass and Pasteur-type pipettes between 1 and 100 ml, and with tips for Sartorius Proline® 5 and 10 ml pipettes.

  • Hydrophobic, autoclavable, 0,45 µm filter prevents over-aspiration
  • Interchangeable adapters for 5 and 10 ml pipette tips
  • Low battery indicator
  • Attached support allows the unit to be rested on a lab bench with the pipette attached
  • Batteries (3×1,2 V) provide approximately seven hours of use when fully charged

The aspiration and dispensing speed can be adjusted by exerting variable pressure on the control buttons.