Anti-Serotonin transporter Rabbit Polyclonal Antibody
Catalog # ANTIA98676-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:5 HTT
- Antigen symbol:Serotonin transporter
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Gene ID:UniprotID# P31645
- Antigen synonyms:5HTT|5 hydroxytryptamine transporter|5HT transporter|SC6A4_HUMAN|hSERT|5 HTTLPR|Na+/Cl- dependent serotonin transporter|5 hydroxytryptamine (serotonin) transporter|OCD1
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
- Molecular weight:70 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human ST (amino acids 348-397)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunising peptide
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Rabbit polyclonal antibody to ST for WB and ELISA with samples derived from human, mouse and rat.
- Validated applications: WB, ELISA
- Recommended dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000
Type: Primary
Antigen: Serotonin transporter
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat