Order Entry
Slovakia
ContactUsLinkComponent
Smart® FFP Masks with Comfortable Nose Seal
  MOLD238501
 :  Moldex
Smart® FFP Masks with Comfortable Nose Seal
  MOLD238501
 :  Moldex
  • Description:
    FFP1 NR D with ventex® valve
  • Environmentally Preferable:
  • Pk:
    20
Smart FFP masks are designed with a soft foam nose cushion for a comfortable fit. A soft, skin-friendly inner fleece layer and a stretchy woven strap provide additional comfort for the wearer.
  • Reliable protection against particles
  • Soft nose cushion for enhanced wearing comfort
  • ActivForm® 3D design for a comfortable and secure fit
The flexible ActivForm® 3D design adapts automatically to the shape of the wearers face and their facial movements. As a result, our Smart masks are easy to wear and sit securely and comfortably on the skin at all times, without the need for a nose clip. Our DuraMesh® structure preserves the shape of the mask under even the most testing conditions.
Smart FFP masks offer reliable protection against particles.
Shelf life from date of production: 5 years.