2,4-Difluoro-5-nitroanisole 96%
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: 1,5-Difluoro-2-methoxy-4-nitrobenzene
CAS Number:179011-39-3
:
Formula:C₇H₅F₂NO₃
MDL Number:MFCD08689675
Molecular Weight:189.1163 g/mol
Melting Pt:93…95 °C
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together